Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605510 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL37 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL37 - interleukin 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014439.3 | 2338 | Intron | NP_055254.2 | |||
NM_173202.1 | 2338 | Intron | NP_775294.1 | |||
NM_173203.1 | 2338 | Intron | NP_775295.1 | |||
NM_173204.1 | 2338 | Intron | NP_775296.1 | |||
NM_173205.1 | 2338 | Intron | NP_775297.1 | |||
XM_011510963.2 | 2338 | UTR 5 | XP_011509265.1 | |||
XM_011510964.2 | 2338 | UTR 5 | XP_011509266.1 |