Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137070 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUS PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FUS - FUS RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170634.1 | 608 | Intron | NP_001164105.1 | |||
NM_001170937.1 | 608 | Intron | NP_001164408.1 | |||
NM_004960.3 | 608 | Intron | NP_004951.1 | |||
XM_005255233.4 | 608 | UTR 5 | XP_005255290.1 | |||
XM_011545781.1 | 608 | Intron | XP_011544083.1 | |||
XM_011545782.2 | 608 | Intron | XP_011544084.1 |
Set Membership: |
HapMap |