Search
Search

CBARP
STK11CCAGGCCATCATCCTGACGTTGGGT[C/T]GGCTGATACACCCCTGTCCTCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBARP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| CBARP - CACN beta subunit associated regulatory protein | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| STK11 - serine/threonine kinase 11 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000455.4 | Intron | NP_000446.1 | ||||
| XM_005259617.2 | Intron | XP_005259674.1 | ||||
| XM_005259618.3 | Intron | XP_005259675.1 | ||||
| XM_011528209.1 | Intron | XP_011526511.1 | ||||