Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATGCTGGAGCCCATGCTGGTCAC[A/G]ATGCTCCCGATGCCAATCCAGAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604024 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC5A6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC5A6 - solute carrier family 5 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021095.2 | 1715 | Silent Mutation | ATC,ATT | I,I 475 | NP_066918.2 | |
XM_006712128.1 | 1715 | Silent Mutation | ATC,ATT | I,I 475 | XP_006712191.1 | |
XM_006712129.1 | 1715 | Silent Mutation | ATC,ATT | I,I 475 | XP_006712192.1 | |
XM_006712130.1 | 1715 | Silent Mutation | ATC,ATT | I,I 475 | XP_006712193.1 | |
XM_011533146.2 | 1715 | Silent Mutation | ATC,ATT | I,I 252 | XP_011531448.1 | |
XM_017005216.1 | 1715 | Silent Mutation | ATC,ATT | I,I 266 | XP_016860705.1 |
Set Membership: |
DME Validated Inventoried |