Search
Search

ASS1CGCAAACAAGTGGAAATCGCCCAGC[G/T]GGAGGGGGCCAAGTATGTGTCCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ASS1 - argininosuccinate synthase 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000050.4 | 679 | Missense Mutation | CGG,CTG | R,L 108 | NP_000041.2 | |
| NM_054012.3 | 679 | Missense Mutation | CGG,CTG | R,L 108 | NP_446464.1 | |
| XM_005272200.3 | 679 | Intron | XP_005272257.1 | |||
| XM_011518705.2 | 679 | Intron | XP_011517007.1 | |||
| XM_017014729.1 | 679 | Intron | XP_016870218.1 | |||