Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TGGCACTCAGCAGTTTCAGGAAGTT[C/T]AGCTGCACGACATTCACTGGGGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120216 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL5A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL5A3 - collagen type V alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015719.3 | 2686 | Silent Mutation | CTA,CTG | L,L 1633 | NP_056534.2 | |
XM_011528042.2 | 2686 | Silent Mutation | CTA,CTG | L,L 1632 | XP_011526344.1 | |
XM_017026849.1 | 2686 | Silent Mutation | CTA,CTG | L,L 854 | XP_016882338.1 |