Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CGGCGACGGCGGGGGCAGCCTGGGA[C/T]TCCACCTCGCGCAGCGCCTCGTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607403 MIM: 612807 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFNL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFNL1 - interferon lambda 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRFN1 - leucine rich repeat and fibronectin type III domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020862.1 | 1836 | Silent Mutation | NP_065913.1 | |||
XM_005259102.4 | 1836 | Missense Mutation | XP_005259159.1 | |||
XM_017027033.1 | 1836 | Missense Mutation | XP_016882522.1 |