Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TGGGTGAAACAGAGTATTAGGGCTC[A/G]GGACTATGTTTGGCTTTGTGCCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300364 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMCX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMCX3 - armadillo repeat containing, X-linked 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ARMCX6 - armadillo repeat containing, X-linked 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009584.1 | Intron | NP_001009584.1 | ||||
NM_001184768.1 | Intron | NP_001171697.1 | ||||
NM_019007.3 | Intron | NP_061880.2 | ||||
XM_011530976.2 | Intron | XP_011529278.1 |