Search
Search

Species: |
Human | ||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||
Phenotype: |
|
||||||||||
Literature Links: |
FCGR3A PubMed Links | ||||||||||
Allele Nomenclature: |
|||||||||||
Minor Allele Frequency: |
| Applied Biosystems® | |||
|---|---|---|---|
Caucasian
|
|||
African American
|
|||
| Chinese - Not Available | |||
| Japanese - Not Available |
| FCGR3A - Fc fragment of IgG receptor IIIa | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001127593.1 | Intron | NP_001121065.1 | ||||
| NM_001127595.1 | Intron | NP_001121067.1 | ||||
| NM_001127596.1 | Intron | NP_001121068.1 | ||||
| XM_011509293.2 | Intron | XP_011507595.2 | ||||
Set Membership: |
|