Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607931 MIM: 608937 MIM: 602389 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATXN2L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATXN2L - ataxin 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4721 - microRNA 4721 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH2B1 - SH2B adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUFM - Tu translation elongation factor, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003321.4 | Intron | NP_003312.3 | ||||
XM_011545928.2 | Intron | XP_011544230.1 | ||||
XM_017023619.1 | Intron | XP_016879108.1 |
Set Membership: |
HapMap |