Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GAGGTGGGGAGCCAAATCGGGAGAC[C/T]GACCTGGGCTCTTGGCCGGAAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611204 MIM: 601404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC88C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC88C - coiled-coil domain containing 88C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080414.3 | 3168 | Missense Mutation | CAG,CGG | Q,R 1984 | NP_001073883.2 | |
XM_005267691.4 | 3168 | Intron | XP_005267748.1 | |||
XM_011536796.2 | 3168 | Missense Mutation | CAG,CGG | Q,R 1948 | XP_011535098.1 | |
XM_017021335.1 | 3168 | Intron | XP_016876824.1 | |||
XM_017021336.1 | 3168 | Missense Mutation | CAG,CGG | Q,R 1011 | XP_016876825.1 | |
XM_017021337.1 | 3168 | Intron | XP_016876826.1 |
GPR68 - G protein-coupled receptor 68 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |