Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| INTS10 - integrator complex subunit 10 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_018142.2 | 281 | Silent Mutation | CGA,CGG | R,R 109 | NP_060612.2 | |
| XM_005273555.3 | 281 | Silent Mutation | CGA,CGG | R,R 109 | XP_005273612.1 | |
| XM_005273556.3 | 281 | Silent Mutation | CGA,CGG | R,R 109 | XP_005273613.1 | |
| XM_005273557.3 | 281 | Silent Mutation | CGA,CGG | R,R 109 | XP_005273614.1 | |
| XM_006716355.2 | 281 | UTR 5 | XP_006716418.1 | |||
| XM_017013606.1 | 281 | Silent Mutation | CGA,CGG | R,R 109 | XP_016869095.1 | |
| XM_017013607.1 | 281 | Silent Mutation | CGA,CGG | R,R 109 | XP_016869096.1 | |
| XM_017013608.1 | 281 | UTR 5 | XP_016869097.1 | |||
| XM_017013609.1 | 281 | UTR 5 | XP_016869098.1 | |||