Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TGCCAACTCTTGCCCTGCTTGGACA[A/G]CAGGTAGACAGCTCCCGGGGAGCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606476 MIM: 182285 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf54 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C19orf54 - chromosome 19 open reading frame 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198476.3 | 1562 | Silent Mutation | CTG,TTG | L,L 288 | NP_940878.3 | |
XM_005258776.4 | 1562 | Silent Mutation | TGC,TGT | C,C 284 | XP_005258833.1 | |
XM_005258778.4 | 1562 | Silent Mutation | CTG,TTG | L,L 116 | XP_005258835.1 | |
XM_006723152.2 | 1562 | Silent Mutation | TGC,TGT | C,C 284 | XP_006723215.1 | |
XM_006723153.2 | 1562 | Silent Mutation | TGC,TGT | C,C 284 | XP_006723216.1 | |
XM_011526774.2 | 1562 | Silent Mutation | CTG,TTG | L,L 288 | XP_011525076.1 | |
XM_011526775.1 | 1562 | Silent Mutation | CTG,TTG | L,L 288 | XP_011525077.1 | |
XM_011526776.2 | 1562 | Silent Mutation | TGC,TGT | C,C 284 | XP_011525078.1 | |
XM_011526777.2 | 1562 | Silent Mutation | CTG,TTG | L,L 288 | XP_011525079.1 | |
XM_011526778.2 | 1562 | Silent Mutation | TGC,TGT | C,C 284 | XP_011525080.1 | |
XM_011526779.2 | 1562 | Silent Mutation | CTG,TTG | L,L 288 | XP_011525081.1 | |
XM_011526782.2 | 1562 | Silent Mutation | CTG,TTG | L,L 288 | XP_011525084.1 | |
XM_011526783.2 | 1562 | Intron | XP_011525085.1 | |||
XM_011526784.2 | 1562 | Silent Mutation | CTG,TTG | L,L 150 | XP_011525086.1 | |
XM_011526785.2 | 1562 | Silent Mutation | CTG,TTG | L,L 116 | XP_011525087.1 | |
XM_011526786.2 | 1562 | Intron | XP_011525088.1 | |||
XM_017026620.1 | 1562 | Silent Mutation | TGC,TGT | C,C 284 | XP_016882109.1 | |
XM_017026621.1 | 1562 | Silent Mutation | CTG,TTG | L,L 288 | XP_016882110.1 | |
XM_017026622.1 | 1562 | Intron | XP_016882111.1 | |||
XM_017026623.1 | 1562 | Intron | XP_016882112.1 | |||
XM_017026624.1 | 1562 | Silent Mutation | CTG,TTG | L,L 150 | XP_016882113.1 | |
XM_017026625.1 | 1562 | Intron | XP_016882114.1 | |||
XM_017026626.1 | 1562 | Silent Mutation | CTG,TTG | L,L 150 | XP_016882115.1 | |
XM_017026627.1 | 1562 | Silent Mutation | CTG,TTG | L,L 116 | XP_016882116.1 |
ITPKC - inositol-trisphosphate 3-kinase C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNRPA - small nuclear ribonucleoprotein polypeptide A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |