Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGAGTCAACACATGTAGACTTGGCA[A/C]AAAGGACAATACAAAAATACAACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf159 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf159 - chromosome 14 open reading frame 159 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPR68 - G protein-coupled receptor 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177676.1 | 2737 | UTR 3 | NP_001171147.1 | |||
NM_003485.3 | 2737 | UTR 3 | NP_003476.3 | |||
XM_005268110.4 | 2737 | UTR 3 | XP_005268167.1 | |||
XM_005268111.3 | 2737 | UTR 3 | XP_005268168.1 | |||
XM_005268112.3 | 2737 | UTR 3 | XP_005268169.1 | |||
XM_006720262.3 | 2737 | UTR 3 | XP_006720325.1 | |||
XM_011537196.2 | 2737 | UTR 3 | XP_011535498.1 | |||
XM_011537197.2 | 2737 | UTR 3 | XP_011535499.1 | |||
XM_011537198.2 | 2737 | UTR 3 | XP_011535500.1 | |||
XM_011537199.2 | 2737 | UTR 3 | XP_011535501.1 |