Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TTCCACCAGCAAGCCAGGGAGGCAG[C/T]ACCACTGGCCCCATTCTGCAGACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606149 MIM: 606150 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FADS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FADS2 - fatty acid desaturase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FADS3 - fatty acid desaturase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021727.4 | Intron | NP_068373.1 | ||||
XM_011545023.2 | Intron | XP_011543325.1 | ||||
XM_017017723.1 | Intron | XP_016873212.1 | ||||
XM_017017724.1 | Intron | XP_016873213.1 |
MIR6746 - microRNA 6746 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |