Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
TTTAGTCAATAATCAAAAATGTTCC[C/T]CAAACGAAGAAGTCCAGGACCAGAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601763 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CASP8 - caspase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080124.1 | Intron | NP_001073593.1 | ||||
NM_001080125.1 | Intron | NP_001073594.1 | ||||
NM_001228.4 | Intron | NP_001219.2 | ||||
NM_033355.3 | Intron | NP_203519.1 | ||||
NM_033356.3 | Intron | NP_203520.1 | ||||
NM_033358.3 | Intron | NP_203522.1 | ||||
XM_005246885.1 | Intron | XP_005246942.1 | ||||
XM_005246886.1 | Intron | XP_005246943.1 | ||||
XM_005246887.1 | Intron | XP_005246944.1 | ||||
XM_005246888.1 | Intron | XP_005246945.1 | ||||
XM_005246889.1 | Intron | XP_005246946.1 | ||||
XM_005246890.3 | Intron | XP_005246947.1 | ||||
XM_005246891.4 | Intron | XP_005246948.1 | ||||
XM_005246892.1 | Intron | XP_005246949.1 | ||||
XM_005246893.2 | Intron | XP_005246950.1 | ||||
XM_005246894.3 | Intron | XP_005246951.1 | ||||
XM_005246895.2 | Intron | XP_005246952.1 | ||||
XM_006712789.1 | Intron | XP_006712852.1 | ||||
XM_006712790.3 | Intron | XP_006712853.1 | ||||
XM_006712793.2 | Intron | XP_006712856.1 | ||||
XM_011511969.1 | Intron | XP_011510271.1 |
Set Membership: |
HapMap |