Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GATCCATCCACAAAGCCAGGCATTA[C/G]AAAAGACAATTTTGGGATGGCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609282 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MOB1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MOB1B - MOB kinase activator 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244766.1 | Intron | NP_001231695.1 | ||||
NM_001244767.1 | Intron | NP_001231696.1 | ||||
NM_173468.3 | Intron | NP_775739.1 | ||||
XM_005265709.2 | Intron | XP_005265766.1 | ||||
XM_011532412.2 | Intron | XP_011530714.1 | ||||
XM_017008837.1 | Intron | XP_016864326.1 |
Set Membership: |
HapMap |