Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C__26338478_10
          See other CEP170 GT Assays ›
          SNP ID:
          rs4512630
          Gene
          CEP170
          Gene Name
          centrosomal protein 170
          Set Membership:
          -
          Chromosome Location:
          Chr.1: 243218516 - 243218516 on Build GRCh38
          Polymorphism:
          C/T, Transition substitution
          Context Sequence [VIC/FAM]:

          TTTCTCAGGCAGAACAACCTCATTT[C/T]TGGTGGCAATTCTGTAAATTGTAGA

          Assay ID C__26338478_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          6 submissions

          Phenotype:

          MIM: 613023

          Literature Links:

          CEP170 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global - Not Available Caucasian - Not Available CEPH (CEU) - Not Available
          EAS - Not Available African American - Not Available YRI (Yoruba) - Not Available
          SAS - Not Available Chinese - Not Available CHB (Han Chinese) - Not Available
          AFR - Not Available Japanese - Not Available JPT (Japanese) - Not Available
          EUR - Not Available
          AMR - Not Available
          CEP170 - centrosomal protein 170
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001042404.1 Intron NP_001035863.1
          NM_001042405.1 Intron NP_001035864.1
          NM_014812.2 Intron NP_055627.2
          XM_006711843.3 Intron XP_006711906.1
          XM_011544334.2 Intron XP_011542636.1
          XM_011544335.2 Intron XP_011542637.1
          XM_011544336.2 Intron XP_011542638.1
          XM_011544337.2 Intron XP_011542639.1
          XM_011544338.2 Intron XP_011542640.1
          XM_011544339.2 Intron XP_011542641.1
          XM_011544340.2 Intron XP_011542642.1
          XM_011544341.2 Intron XP_011542643.1
          XM_011544342.2 Intron XP_011542644.1
          XM_011544343.2 Intron XP_011542645.1
          XM_011544344.2 Intron XP_011542646.1
          XM_017002932.1 Intron XP_016858421.1
          XM_017002933.1 Intron XP_016858422.1
          XM_017002934.1 Intron XP_016858423.1
          XM_017002935.1 Intron XP_016858424.1
          XM_017002936.1 Intron XP_016858425.1
          XM_017002937.1 Intron XP_016858426.1
          XM_017002938.1 Intron XP_016858427.1
          XM_017002939.1 Intron XP_016858428.1
          XM_017002940.1 Intron XP_016858429.1
          XM_017002941.1 Intron XP_016858430.1
          XM_017002942.1 Intron XP_016858431.1
          XM_017002943.1 Intron XP_016858432.1
          XM_017002944.1 Intron XP_016858433.1
          XM_017002945.1 Intron XP_016858434.1
          XM_017002946.1 Intron XP_016858435.1
          XM_017002947.1 Intron XP_016858436.1
          XM_017002948.1 Intron XP_016858437.1
          XM_017002949.1 Intron XP_016858438.1
          XM_017002950.1 Intron XP_016858439.1
          XM_017002951.1 Intron XP_016858440.1

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline