Search
Search

CIDECGCAGGTAACAGGAGGTGCCAAGCAG[C/T]ACGTGGCCTGTGGCCTGCATGCTGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||
Literature Links: |
CIDEC PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
| EUR - Not Available | |||||
| AMR - Not Available |
| CIDEC - cell death inducing DFFA like effector c | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001199551.1 | 689 | Silent Mutation | GTA,GTG | V,V 211 | NP_001186480.1 | |
| NM_001199552.1 | 689 | Silent Mutation | GTA,GTG | V,V 201 | NP_001186481.1 | |
| NM_001199623.1 | 689 | Silent Mutation | GTA,GTG | V,V 214 | NP_001186552.1 | |
| NM_001321142.1 | 689 | Silent Mutation | GTA,GTG | V,V 201 | NP_001308071.1 | |
| NM_001321143.1 | 689 | Silent Mutation | GTA,GTG | V,V 127 | NP_001308072.1 | |
| NM_001321144.1 | 689 | Silent Mutation | GTA,GTG | V,V 127 | NP_001308073.1 | |
| NM_022094.3 | 689 | Silent Mutation | GTA,GTG | V,V 201 | NP_071377.2 | |