Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 147880 MIM: 610482 | ||||||||||||||||||||
Literature Links: |
IL6R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL6R - interleukin 6 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000565.3 | 1903 | Intron | NP_000556.1 | |||
NM_001206866.1 | 1903 | Intron | NP_001193795.1 | |||
NM_181359.2 | 1903 | Intron | NP_852004.1 | |||
XM_005245139.1 | 1903 | Intron | XP_005245196.1 | |||
XM_005245140.2 | 1903 | Intron | XP_005245197.1 | |||
XM_006711298.1 | 1903 | Intron | XP_006711361.1 | |||
XM_006711299.3 | 1903 | Intron | XP_006711362.1 | |||
XM_017001199.1 | 1903 | Intron | XP_016856688.1 | |||
XM_017001200.1 | 1903 | Intron | XP_016856689.1 | |||
XM_017001201.1 | 1903 | Intron | XP_016856690.1 |
SHE - Src homology 2 domain containing E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010846.2 | 1903 | Intron | NP_001010846.1 | |||
XM_005244891.4 | 1903 | UTR 3 | XP_005244948.1 | |||
XM_011509163.2 | 1903 | UTR 3 | XP_011507465.1 | |||
XM_017000256.1 | 1903 | Intron | XP_016855745.1 |
Set Membership: |
JSNP |