Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608880 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFYVE16 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ZFYVE16 - zinc finger FYVE-type containing 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105251.2 | 10 | UTR 5 | NP_001098721.1 | |||
NM_001284236.1 | 10 | UTR 5 | NP_001271165.1 | |||
NM_001284237.1 | 10 | UTR 5 | NP_001271166.1 | |||
NM_014733.4 | 10 | UTR 5 | NP_055548.3 | |||
XM_005248632.4 | 10 | Intron | XP_005248689.1 | |||
XM_011543753.2 | 10 | Intron | XP_011542055.1 | |||
XM_017010091.1 | 10 | UTR 5 | XP_016865580.1 | |||
XM_017010092.1 | 10 | Intron | XP_016865581.1 | |||
XM_017010093.1 | 10 | Intron | XP_016865582.1 | |||
XM_017010094.1 | 10 | Intron | XP_016865583.1 | |||
XM_017010095.1 | 10 | Intron | XP_016865584.1 | |||
XM_017010096.1 | 10 | Intron | XP_016865585.1 |