Search
Search

APOC1
APOE
TOMM40CCAGAGCACCGAGGAGCTGCGGGTG[C/A]GCCTCGCCTCCCACCTGCGCAAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
APOC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| APOC1 - apolipoprotein C1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| APOE - apolipoprotein E | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000041.3 | 658 | Missense Mutation | AGC,CGC | S,R 154 | NP_000032.1 | |
| NM_001302688.1 | 658 | Missense Mutation | AGC,CGC | S,R 180 | NP_001289617.1 | |
| NM_001302689.1 | 658 | Missense Mutation | AGC,CGC | S,R 154 | NP_001289618.1 | |
| NM_001302690.1 | 658 | Missense Mutation | AGC,CGC | S,R 154 | NP_001289619.1 | |
| NM_001302691.1 | 658 | Missense Mutation | AGC,CGC | S,R 154 | NP_001289620.1 | |
| TOMM40 - translocase of outer mitochondrial membrane 40 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||