Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
GGGCCGCATAAGGAAGGAACTAGTG[C/T]GGAGGTCCGGGGAGCGGCCGCGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607112 MIM: 609111 MIM: 605647 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO2 - F-box protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXO44 - F-box protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014765.1 | 12 | Intron | NP_001014765.1 | |||
NM_001304790.1 | 12 | Intron | NP_001291719.1 | |||
NM_001304791.1 | 12 | Intron | NP_001291720.1 | |||
NM_033182.5 | 12 | Intron | NP_149438.2 | |||
NM_183412.2 | 12 | Intron | NP_904319.1 | |||
NM_183413.2 | 12 | Intron | NP_904320.1 | |||
XM_005263535.1 | 12 | Intron | XP_005263592.1 | |||
XM_005263536.4 | 12 | Intron | XP_005263593.1 | |||
XM_005263537.1 | 12 | Intron | XP_005263594.1 | |||
XM_006711043.2 | 12 | Intron | XP_006711106.1 | |||
XM_006711045.2 | 12 | Intron | XP_006711108.1 | |||
XM_011542435.1 | 12 | Intron | XP_011540737.1 | |||
XM_017002842.1 | 12 | Intron | XP_016858331.1 | |||
XM_017002843.1 | 12 | Intron | XP_016858332.1 | |||
XM_017002844.1 | 12 | Intron | XP_016858333.1 |
FBXO6 - F-box protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018438.5 | 12 | UTR 5 | NP_060908.1 | |||
XM_005263448.4 | 12 | Intron | XP_005263505.1 | |||
XM_005263449.4 | 12 | Intron | XP_005263506.1 | |||
XM_005263451.4 | 12 | Intron | XP_005263508.1 | |||
XM_006710571.3 | 12 | Intron | XP_006710634.1 | |||
XM_011541232.2 | 12 | Intron | XP_011539534.1 | |||
XM_017001007.1 | 12 | Intron | XP_016856496.1 |