Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMYD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| SMYD3 - SET and MYND domain containing 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001167740.1 | Intron | NP_001161212.1 | ||||
| NM_022743.2 | Intron | NP_073580.1 | ||||
| XM_011544253.1 | Intron | XP_011542555.1 | ||||
| XM_011544254.2 | Intron | XP_011542556.1 | ||||
| XM_011544257.1 | Intron | XP_011542559.1 | ||||
| XM_011544258.2 | Intron | XP_011542560.1 | ||||
| XM_011544259.2 | Intron | XP_011542561.1 | ||||
| XM_011544260.1 | Intron | XP_011542562.1 | ||||
| XM_011544261.2 | Intron | XP_011542563.1 | ||||
| XM_017002094.1 | Intron | XP_016857583.1 | ||||
| XM_017002095.1 | Intron | XP_016857584.1 | ||||