Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD31 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ANKRD31 - ankyrin repeat domain 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164443.1 | Intron | NP_001157915.1 | ||||
XM_011543298.2 | Intron | XP_011541600.1 | ||||
XM_011543299.1 | Intron | XP_011541601.1 | ||||
XM_011543300.2 | Intron | XP_011541602.1 | ||||
XM_011543301.2 | Intron | XP_011541603.1 | ||||
XM_011543302.2 | Intron | XP_011541604.1 | ||||
XM_017009317.1 | Intron | XP_016864806.1 | ||||
XM_017009318.1 | Intron | XP_016864807.1 | ||||
XM_017009319.1 | Intron | XP_016864808.1 | ||||
XM_017009320.1 | Intron | XP_016864809.1 | ||||
XM_017009321.1 | Intron | XP_016864810.1 | ||||
XM_017009322.1 | Intron | XP_016864811.1 |