Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
62 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609121 MIM: 603272 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATSPER4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CATSPER4 - cation channel sperm associated 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198137.1 | 393 | Intron | NP_937770.1 | |||
XM_011541432.2 | 393 | Intron | XP_011539734.1 | |||
XM_011541433.2 | 393 | UTR 5 | XP_011539735.1 |
CNKSR1 - connector enhancer of kinase suppressor of Ras 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297647.1 | 393 | Intron | NP_001284576.1 | |||
NM_001297648.1 | 393 | Intron | NP_001284577.1 | |||
NM_006314.2 | 393 | Intron | NP_006305.2 | |||
XM_011540484.2 | 393 | Intron | XP_011538786.1 | |||
XM_011540485.2 | 393 | Intron | XP_011538787.1 |