Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
67 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf167 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| C1orf167 - chromosome 1 open reading frame 167 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| MTHFR - methylenetetrahydrofolate reductase (NAD(P)H) | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_005957.4 | Intron | NP_005948.3 | ||||
| XM_005263458.3 | Intron | XP_005263515.1 | ||||
| XM_005263460.4 | Intron | XP_005263517.1 | ||||
| XM_005263462.4 | Intron | XP_005263519.1 | ||||
| XM_005263463.3 | Intron | XP_005263520.1 | ||||
| XM_011541495.2 | Intron | XP_011539797.1 | ||||
| XM_011541496.2 | Intron | XP_011539798.1 | ||||
| XM_017001328.1 | Intron | XP_016856817.1 | ||||