Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606439 MIM: 604923 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATL1 - atlastin GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127713.1 | Intron | NP_001121185.1 | ||||
NM_015915.4 | Intron | NP_056999.2 | ||||
NM_181598.3 | Intron | NP_853629.2 |
MAP4K5 - mitogen-activated protein kinase kinase kinase kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006575.4 | Intron | NP_006566.2 | ||||
NM_198794.2 | Intron | NP_942089.1 | ||||
XM_006720013.3 | Intron | XP_006720076.1 | ||||
XM_011536377.2 | Intron | XP_011534679.1 | ||||
XM_011536378.2 | Intron | XP_011534680.1 | ||||
XM_011536379.2 | Intron | XP_011534681.1 | ||||
XM_011536380.2 | Intron | XP_011534682.1 |
Set Membership: |
HapMap |