Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610732 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TTC12 - tetratricopeptide repeat domain 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318533.1 | Intron | NP_001305462.1 | ||||
NM_017868.3 | Intron | NP_060338.3 | ||||
XM_005271604.3 | Intron | XP_005271661.1 | ||||
XM_011542890.2 | Intron | XP_011541192.1 | ||||
XM_017017943.1 | Intron | XP_016873432.1 | ||||
XM_017017944.1 | Intron | XP_016873433.1 | ||||
XM_017017945.1 | Intron | XP_016873434.1 | ||||
XM_017017946.1 | Intron | XP_016873435.1 | ||||
XM_017017947.1 | Intron | XP_016873436.1 | ||||
XM_017017948.1 | Intron | XP_016873437.1 | ||||
XM_017017949.1 | Intron | XP_016873438.1 | ||||
XM_017017950.1 | Intron | XP_016873439.1 |