Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| ABCC4 - ATP binding cassette subfamily C member 4 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001105515.2 | Intron | NP_001098985.1 | ||||
| NM_001301829.1 | Intron | NP_001288758.1 | ||||
| NM_001301830.1 | Intron | NP_001288759.1 | ||||
| NM_005845.4 | Intron | NP_005836.2 | ||||
| XM_005254025.2 | Intron | XP_005254082.1 | ||||
| XM_017020319.1 | Intron | XP_016875808.1 | ||||
| XM_017020320.1 | Intron | XP_016875809.1 | ||||
| XM_017020321.1 | Intron | XP_016875810.1 | ||||
| XM_017020322.1 | Intron | XP_016875811.1 | ||||
Set Membership: |
|