Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| AK7 - adenylate kinase 7 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_152327.3 | Intron | NP_689540.2 | ||||
| XM_006720021.2 | Intron | XP_006720084.1 | ||||
| XM_006720022.2 | Intron | XP_006720085.1 | ||||
| XM_011536403.2 | Intron | XP_011534705.1 | ||||
| XM_017020956.1 | Intron | XP_016876445.1 | ||||
| XM_017020957.1 | Intron | XP_016876446.1 | ||||
| XM_017020958.1 | Intron | XP_016876447.1 | ||||
| XM_017020959.1 | Intron | XP_016876448.1 | ||||
| XM_017020960.1 | Intron | XP_016876449.1 | ||||