Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCCGTTAAGGAAGCAGCACATC[A/G]CTGCATTCGGCTGGTTTTCAGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608086 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIMAP1-GIMAP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GIMAP1-GIMAP5 - GIMAP1-GIMAP5 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199577.1 | 37 | Intron | NP_001186506.1 | |||
NM_001303630.1 | 37 | Intron | NP_001290559.1 |
GIMAP5 - GTPase, IMAP family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018384.4 | 37 | UTR 5 | NP_060854.2 |
Set Membership: |
HapMap |