Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600275 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOTCH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOTCH2 - notch 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001200001.1 | 8141 | Intron | NP_001186930.1 | |||
NM_024408.3 | 8141 | UTR 3 | NP_077719.2 | |||
XM_005270901.3 | 8141 | UTR 3 | XP_005270958.1 | |||
XM_011541519.2 | 8141 | UTR 3 | XP_011539821.1 | |||
XM_011541520.2 | 8141 | UTR 3 | XP_011539822.1 | |||
XM_017001372.1 | 8141 | UTR 3 | XP_016856861.1 | |||
XM_017001373.1 | 8141 | UTR 3 | XP_016856862.1 |