Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600970 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MYO6 - myosin VI | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300899.1 | Intron | NP_001287828.1 | ||||
NM_004999.3 | Intron | NP_004990.3 | ||||
XM_005248719.3 | Intron | XP_005248776.1 | ||||
XM_005248720.3 | Intron | XP_005248777.1 | ||||
XM_005248721.3 | Intron | XP_005248778.1 | ||||
XM_005248722.3 | Intron | XP_005248779.1 | ||||
XM_005248724.3 | Intron | XP_005248781.1 | ||||
XM_005248726.3 | Intron | XP_005248783.1 | ||||
XM_017010899.1 | Intron | XP_016866388.1 |
Set Membership: |
HapMap |