Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C__30044419_20
          See other LINC00473 GT Assays ›
          SNP ID:
          rs9356405
          Gene
          LINC00473 PDE10A
          Gene Name
          long intergenic non-protein coding RNA 473
          phosphodiesterase 10A
          Set Membership:
          > HapMap
          Chromosome Location:
          Chr.6: 165930591 - 165930591 on Build GRCh38
          Polymorphism:
          G/T, Transversion Substitution
          Context Sequence [VIC/FAM]:

          TTGTTTTCTTCAAAAGATACTCAGG[G/T]TCTGCCTTCATCAGTATTCCAATAA

          Assay ID C__30044419_20
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 610652

          Literature Links:

          LINC00473 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          T (0.21)
          (0.79)
          Caucasian - Not Available CEPH (CEU)
          T (0.21)
          (0.79)
          EAS
          T (0.28)
          (0.72)
          African American - Not Available YRI (Yoruba)
          T (0.22)
          (0.78)
          SAS
          T (0.09)
          (0.91)
          Chinese - Not Available JPT (Japanese)
          T (0.30)
          (0.70)
          AFR
          T (0.22)
          (0.78)
          Japanese - Not Available CHB (Han Chinese)
          T (0.26)
          (0.74)
          EUR
          T (0.18)
          (0.82)
          AMR
          T (0.29)
          (0.71)
          LINC00473 - long intergenic non-protein coding RNA 473
          There are no transcripts associated with this gene.
          PDE10A - phosphodiesterase 10A
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001130690.2 Intron NP_001124162.1
          NM_006661.3 Intron NP_006652.1
          XM_006715321.3 Intron XP_006715384.1
          XM_011535387.2 Intron XP_011533689.2
          XM_011535388.2 Intron XP_011533690.1
          XM_011535393.2 Intron XP_011533695.1
          XM_017010194.1 Intron XP_016865683.1
          XM_017010195.1 Intron XP_016865684.1
          XM_017010196.1 Intron XP_016865685.1
          XM_017010197.1 Intron XP_016865686.1

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs77343208] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          HapMap

          Panther Classification:

          Molecular Function -

          hydrolase phosphodiesterase

          Gene Ontology Categories:

          Function(s) Process(es)

          cAMP catabolic process
          signal transduction
          regulation of protein kinase A signaling
          regulation of cAMP-mediated signaling
          cGMP catabolic process
          3',5'-cyclic-nucleotide phosphodiesterase activity
          3',5'-cyclic-AMP phosphodiesterase activity
          cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
          drug binding
          cAMP binding
          cGMP binding
          metal ion binding
          3',5'-cyclic-GMP phosphodiesterase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline