Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIGN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| PIGN - phosphatidylinositol glycan anchor biosynthesis class N | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_012327.5 | Intron | NP_036459.1 | ||||
| NM_176787.4 | Intron | NP_789744.1 | ||||
| XM_011525889.1 | Intron | XP_011524191.1 | ||||
| XM_011525890.1 | Intron | XP_011524192.1 | ||||
| XM_011525891.1 | Intron | XP_011524193.1 | ||||
| XM_011525892.1 | Intron | XP_011524194.1 | ||||
| XM_011525893.1 | Intron | XP_011524195.1 | ||||
| XM_011525894.1 | Intron | XP_011524196.1 | ||||
| XM_011525895.1 | Intron | XP_011524197.1 | ||||
| XM_011525896.1 | Intron | XP_011524198.1 | ||||
| XM_011525898.1 | Intron | XP_011524200.1 | ||||
| XM_017025685.1 | Intron | XP_016881174.1 | ||||
| XM_017025686.1 | Intron | XP_016881175.1 | ||||