Search
Search

Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||
Literature Links: |
MITF PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
| EUR - Not Available | |||||
| AMR - Not Available |
| MITF - melanogenesis associated transcription factor | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000248.3 | Intron | NP_000239.1 | ||||
| NM_001184967.1 | Intron | NP_001171896.1 | ||||
| NM_001184968.1 | Intron | NP_001171897.1 | ||||
| NM_006722.2 | Intron | NP_006713.1 | ||||
| NM_198158.2 | Intron | NP_937801.1 | ||||
| NM_198159.2 | Intron | NP_937802.1 | ||||
| NM_198177.2 | Intron | NP_937820.1 | ||||
| NM_198178.2 | Intron | NP_937821.2 | ||||
| XM_005264754.1 | Intron | XP_005264811.1 | ||||
| XM_005264755.3 | Intron | XP_005264812.1 | ||||
| XM_006713164.2 | Intron | XP_006713227.1 | ||||
| XM_011533722.2 | Intron | XP_011532024.1 | ||||
| XM_011533723.2 | Intron | XP_011532025.1 | ||||
| XM_011533725.1 | Intron | XP_011532027.1 | ||||
| XM_011533726.1 | Intron | XP_011532028.1 | ||||
| XM_017006444.1 | Intron | XP_016861933.1 | ||||
| XM_017006445.1 | Intron | XP_016861934.1 | ||||
| XM_017006446.1 | Intron | XP_016861935.1 | ||||
| XM_017006447.1 | Intron | XP_016861936.1 | ||||
| XM_017006448.1 | Intron | XP_016861937.1 | ||||