Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ADGRL3 - adhesion G protein-coupled receptor L3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001322246.1 | Intron | NP_001309175.1 | ||||
| NM_001322402.1 | Intron | NP_001309331.1 | ||||
| NM_015236.5 | Intron | NP_056051.2 | ||||
| XM_011531788.2 | Intron | XP_011530090.1 | ||||
| XM_011531791.2 | Intron | XP_011530093.1 | ||||
| XM_017007929.1 | Intron | XP_016863418.1 | ||||
| XM_017007930.1 | Intron | XP_016863419.1 | ||||
| XM_017007931.1 | Intron | XP_016863420.1 | ||||
| XM_017007932.1 | Intron | XP_016863421.1 | ||||
| XM_017007933.1 | Intron | XP_016863422.1 | ||||
| XM_017007934.1 | Intron | XP_016863423.1 | ||||
| XM_017007935.1 | Intron | XP_016863424.1 | ||||
| XM_017007936.1 | Intron | XP_016863425.1 | ||||
| XM_017007937.1 | Intron | XP_016863426.1 | ||||
| XM_017007938.1 | Intron | XP_016863427.1 | ||||
| XM_017007939.1 | Intron | XP_016863428.1 | ||||
| XM_017007940.1 | Intron | XP_016863429.1 | ||||
| XM_017007941.1 | Intron | XP_016863430.1 | ||||
| XM_017007942.1 | Intron | XP_016863431.1 | ||||