Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATACATATTTCATGACTAACTTT[C/T]GGGTTTTGGGTACTTTTTTTAACCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611730 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPB41L5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EPB41L5 - erythrocyte membrane protein band 4.1 like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184937.1 | Intron | NP_001171866.1 | ||||
NM_020909.3 | Intron | NP_065960.2 | ||||
XM_006712651.1 | Intron | XP_006712714.1 | ||||
XM_006712652.1 | Intron | XP_006712715.1 | ||||
XM_006712653.1 | Intron | XP_006712716.1 | ||||
XM_011511529.2 | Intron | XP_011509831.1 | ||||
XM_017004567.1 | Intron | XP_016860056.1 | ||||
XM_017004568.1 | Intron | XP_016860057.1 |