Search
Search

DHPS
MAN2B1
WDR83
WDR83OSTACTTCATACCTAGATGCCTGCCCC[A/G]TCTCAGCCCTGTCCTTACCTCAGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHPS PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| DHPS - deoxyhypusine synthase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001206974.1 | Intron | NP_001193903.1 | ||||
| NM_001930.3 | Intron | NP_001921.1 | ||||
| NM_013406.2 | Intron | NP_037538.1 | ||||
| XM_011527770.1 | Intron | XP_011526072.1 | ||||
| XM_011527771.2 | Intron | XP_011526073.1 | ||||
| MAN2B1 - mannosidase alpha class 2B member 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| WDR83 - WD repeat domain 83 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001099737.2 | Intron | NP_001093207.1 | ||||
| NM_032332.3 | Intron | NP_115708.1 | ||||
| WDR83OS - WD repeat domain 83 opposite strand | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||