Search
Search

Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MASP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| MASP2 - mannan binding lectin serine peptidase 2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| TARDBP - TAR DNA binding protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_007375.3 | Intron | NP_031401.1 | ||||
| XM_017000863.1 | Intron | XP_016856352.1 | ||||
| XM_017000864.1 | Intron | XP_016856353.1 | ||||
| XM_017000865.1 | Intron | XP_016856354.1 | ||||
| XM_017000866.1 | Intron | XP_016856355.1 | ||||
| XM_017000867.1 | Intron | XP_016856356.1 | ||||
| XM_017000868.1 | Intron | XP_016856357.1 | ||||