Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C__30567694_10
          See other LOC101927139 GT Assays ›
          SNP ID:
          rs7530150
          Gene
          LOC101927139 PDE4B
          Gene Name
          uncharacterized LOC101927139
          phosphodiesterase 4B
          Set Membership:
          > HapMap
          Chromosome Location:
          Chr.1: 66047991 - 66047991 on Build GRCh38
          Polymorphism:
          A/G, Transition substitution
          Context Sequence [VIC/FAM]:

          TCATTTCCACAGAGCATGCATACAC[A/G]TTCTTTCTTTGGATGGCGTCTACAG

          Assay ID C__30567694_10
          Size
          Availability Made To Order
          Catalog # 4351379
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          46 submissions

          Phenotype:

          MIM: 600127

          Literature Links:

          LOC101927139 PubMed Links

          Allele Nomenclature:

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global
          A (0.14)
          (0.86)
          Caucasian - Not Available CEPH (CEU)
          A (0.01)
          (0.99)
          EAS
          A (0.35)
          (0.65)
          African American - Not Available YRI (Yoruba)
          A (0.11)
          (0.89)
          SAS
          A (0.21)
          (0.79)
          Chinese - Not Available JPT (Japanese)
          A (0.29)
          (0.71)
          AFR
          A (0.08)
          (0.92)
          Japanese - Not Available CHB (Han Chinese)
          A (0.36)
          (0.64)
          EUR
          A (0.01)
          (0.99)
          AMR
          A (0.07)
          (0.93)
          LOC101927139 - uncharacterized LOC101927139
          There are no transcripts associated with this gene.
          PDE4B - phosphodiesterase 4B
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_001037339.2 Intron NP_001032416.1
          NM_001037340.2 Intron NP_001032417.1
          NM_001037341.1 Intron NP_001032418.1
          NM_001297440.1 Intron NP_001284369.1
          NM_001297441.1 Intron NP_001284370.1
          NM_001297442.1 Intron NP_001284371.1
          NM_002600.3 Intron NP_002591.2
          XM_005270924.3 Intron XP_005270981.1
          XM_005270925.2 Intron XP_005270982.1
          XM_006710680.3 Intron XP_006710743.1
          XM_011541566.1 Intron XP_011539868.1
          XM_017001445.1 Intron XP_016856934.1
          XM_017001446.1 Intron XP_016856935.1

          Back To Top

          More Information


          Set Membership:

          HapMap

          Panther Classification:

          Molecular Function -

          phosphodiesterase

          Gene Ontology Categories:

          Function(s) Process(es)

          neutrophil homeostasis
          cAMP catabolic process
          neutrophil chemotaxis
          positive regulation of interferon-gamma production
          positive regulation of interleukin-2 production
          cellular response to drug
          T cell receptor signaling pathway
          leukocyte migration
          cellular response to lipopolysaccharide
          cellular response to epinephrine stimulus
          regulation of cardiac muscle cell contraction
          regulation of high voltage-gated calcium channel activity
          negative regulation of relaxation of cardiac muscle
          3',5'-cyclic-AMP phosphodiesterase activity
          cAMP binding
          gamma-tubulin binding
          ion channel binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline