Search Thermo Fisher Scientific
GGCCTGGAGGCAGGGTGGGTAGACA[A/G]GATGACCATTGACCTCATGCAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607368 MIM: 611601 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNK15 - potassium two pore domain channel subfamily K member 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNK15-AS1 - KCNK15 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIMS4 - regulating synaptic membrane exocytosis 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205317.1 | 1820 | UTR 3 | NP_001192246.1 | |||
NM_182970.3 | 1820 | UTR 3 | NP_892015.1 |
Set Membership: |
HapMap |