Search
Search

BLOC1S3
TRAPPC6AAAGGTGGCATTTCCTGCACACCCCT[C/G]CAGGCCAAGATGATGCTCGCTCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
BLOC1S3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| BLOC1S3 - biogenesis of lysosomal organelles complex 1 subunit 3 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_212550.4 | 2158 | UTR 3 | NP_997715.1 | |||
| TRAPPC6A - trafficking protein particle complex 6A | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||