Search
Search

SLC22A1GACATTCTGGAGCAGGTTGGGGAGT[C/T]TGGCTGGTTCCAGAAGCAAGCCTTC
Species: |
Human | ||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||
Phenotype: |
|
||||||||||||||||
Literature Links: |
SLC22A1 PubMed Links | ||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||
Minor Allele Frequency: |
| Applied Biosystems® | |||
|---|---|---|---|
Caucasian
|
|||
African American
|
|||
Japanese
|
|||
Chinese
|
| SLC22A1 - solute carrier family 22 member 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_003057.2 | 146 | Missense Mutation | TCT,TTT | S,F 14 | NP_003048.1 | |
| NM_153187.1 | 146 | Missense Mutation | TCT,TTT | S,F 14 | NP_694857.1 | |
| XM_005267102.4 | 188 | Missense Mutation | TCT,TTT | S,F 14 | XP_005267159.1 | |
| XM_005267103.1 | 188 | Missense Mutation | TCT,TTT | S,F 14 | XP_005267160.1 | |
| XM_005267104.4 | Intron | XP_005267161.1 | ||||
| XM_005267105.4 | Intron | XP_005267162.1 | ||||
| XM_006715552.1 | 188 | Missense Mutation | TCT,TTT | S,F 14 | XP_006715615.1 | |
| XM_011536074.2 | Intron | XP_011534376.1 | ||||
Set Membership: |
|