Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › C__30634131_20
          See other CYP2C9 GT Assays ›
          SNP ID:
          rs56165452
          Gene
          CYP2C9
          Gene Name
          cytochrome P450 family 2 subfamily C member 9
          Set Membership:
          > DME > Validated > Inventoried
          Chromosome Location:
          Chr.10: 94981297 - 94981297 on Build GRCh38
          Polymorphism:
          C/T, Transition substitution
          Context Sequence [VIC/FAM]:

          GTGGTGCACGAGGTCCAGAGATACA[C/T]TGACCTTCTCCCCACCAGCCTGCCC

          Assay ID C__30634131_20
          Size 150 rxns
          Availability Inventoried
          Catalog # 4362691
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details



          Species:

          Human

          dbSNP Submissions:

          NA

          Phenotype:

          MIM: 601130

          Literature Links:

          CYP2C9 PubMed Links

          Allele Nomenclature:

          CYP2C9*4,c.1076T>C CYP2C9*4,g.42615T>C

          Minor Allele Frequency:

          1000Genome Applied Biosystems® HapMap
          Global - Not Available Caucasian
          C (0.00)
          (1.00)
          CEPH (CEU) - Not Available
          EAS - Not Available African American
          C (0.00)
          (1.00)
          YRI (Yoruba) - Not Available
          SAS - Not Available Japanese
          C (0.00)
          (1.00)
          CHB (Han Chinese) - Not Available
          AFR - Not Available Chinese
          C (0.00)
          (1.00)
          JPT (Japanese) - Not Available
          EUR - Not Available
          AMR - Not Available
          CYP2C9 - cytochrome P450 family 2 subfamily C member 9
          Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
          NM_000771.3 1101 Missense Mutation ACT,ATT T,I 359 NP_000762.2
          XM_017015758.1 1101 Missense Mutation ACT,ATT T,I 359 XP_016871247.1

          Back To Top

          More Information


          Important Information

          Assay C__30634131_20 detects the rare CYP2C9*4 c.1076T>C SNP, which is adjacent to the polymorphic CYP2C9*3,*18 c.1075A>C SNP that is detected by assay C__27104892_10. These two SNP assays can be used to detect the 3 possible haplotypes described for this locus (www.pharmvar.org/gene/CYP2C9; minor alleles 1076C and 1075C do not occur together in a haplotype). Run both assays separately on the same samples and analyze data as described in the PGx Experiments User Guide (Pub. # MAN0009612) Chapter 5 section 'TaqMan DME genotyping assays to triallelic SNPs and adjacent SNP targets'.
          Assay C__27104892_10 interrogates the CYP2C9*3,*18 c.1075A>C SNP (rs1057910).

          Additional Information:

          For this assay, SNP(s) [rs1057910] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance. Assay C__27104892_10 interrogates rs1057910 (CYP2C9*3/*18 c.1075A>C). For more information, refer to the assay Important Information note.

          Set Membership:

          DME Validated Inventoried

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          steroid metabolic process
          monoterpenoid metabolic process
          drug metabolic process
          epoxygenase P450 pathway
          urea metabolic process
          monocarboxylic acid metabolic process
          drug catabolic process
          exogenous drug catabolic process
          cellular amide metabolic process
          oxidation-reduction process
          oxidative demethylation
          omega-hydroxylase P450 pathway
          cholesterol 25-hydroxylase activity
          monooxygenase activity
          iron ion binding
          drug binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity
          (S)-limonene 6-monooxygenase activity
          (S)-limonene 7-monooxygenase activity
          oxygen binding
          heme binding
          caffeine oxidase activity
          (R)-limonene 6-monooxygenase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline