Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
ACTTTCATATGTTCAAATCCCATGG[A/T]GGAGTGTTTCATCCTAGAAACTCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605097 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC45A3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC45A3 - solute carrier family 45 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033102.2 | 2643 | UTR 3 | NP_149093.1 | |||
XM_005245556.3 | 2643 | UTR 3 | XP_005245613.1 | |||
XM_005245557.4 | 2643 | UTR 3 | XP_005245614.1 | |||
XM_005245560.2 | 2643 | Intron | XP_005245617.1 | |||
XM_017002638.1 | 2643 | UTR 3 | XP_016858127.1 |
Set Membership: |
HapMap |