Search
Search

MVB12BTGTTTCAGAGCCCATCTGCTCCTTG[C/T]AGTTCCCCGAGCAGTCCCTGTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MVB12B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| MVB12B - multivesicular body subunit 12B | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001011703.2 | Intron | NP_001011703.1 | ||||
| NM_033446.2 | Intron | NP_258257.1 | ||||
| XM_005252297.1 | Intron | XP_005252354.1 | ||||
| XM_017015275.1 | Intron | XP_016870764.1 | ||||
| XM_017015276.1 | Intron | XP_016870765.1 | ||||
| XM_017015277.1 | Intron | XP_016870766.1 | ||||
| XM_017015278.1 | Intron | XP_016870767.1 | ||||
| XM_017015279.1 | Intron | XP_016870768.1 | ||||