Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAATTTTGAATATTCTAAATTTA[G/T]TAATTGGCAGGAAAGACCATGTGGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611193 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRP15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RRP15 - ribosomal RNA processing 15 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016052.3 | Intron | NP_057136.2 | ||||
XM_011509597.2 | Intron | XP_011507899.1 |