Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
GTGGTTTGGTTGCCAGGATATCTCT[C/G]GACAGGGCCCTTGACAGCCACTGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602495 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL24 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCL24 - C-C motif chemokine ligand 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002991.2 | 595 | Missense Mutation | CAG,GAG | Q,E 110 | NP_002982.2 | |
XM_011516459.2 | 595 | Missense Mutation | CAG,GAG | Q,E 110 | XP_011514761.1 | |
XM_011516460.2 | 595 | Missense Mutation | CAG,GAG | Q,E 110 | XP_011514762.1 |
Set Membership: |
HapMap |